Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UBR5/EDD Antibody, Novus Biologicals™
SDP

Catalog No. NB1001220 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1001220 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1001220 Supplier Novus Biologicals Supplier No. NB1001220
Add to Cart
Add to Cart

Goat Polyclonal Antibody has been used in 2 publications

UBR5/EDD Polyclonal specifically detects UBR5/EDD in Human, Mouse, Rat, Rabbit samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.

Specifications

Antigen UBR5/EDD
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Immunohistochemistry 1 - 2 ug/ml, Immunohistochemistry-Paraffin 1 - 2 ug/ml, Peptide ELISA Detection limit 1:4000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_056986.
Gene Alias DD5, E3 ubiquitin protein ligase, HECT domain containing, 1, E3 ubiquitin-protein ligase, HECT domain-containing 1, EC 6.3.2, EC 6.3.2.-, EDD1E3 ubiquitin-protein ligase UBR5, EDDE3 identified by differential display, hHYD, HYDFLJ11310, Hyperplastic discs protein homolog, KIAA0896MGC57263, progestin induced protein, Progestin-induced protein, ubiquitin protein ligase E3 component n-recognin 5, ubiquitin-protein ligase
Gene Symbols UBR5
Host Species Goat
Immunogen Peptide with sequence C-LAIKTKNFGFV corresponding to C-Terminus according to NP_056986.2.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline DNA Repair
Primary or Secondary Primary
Gene ID (Entrez) 51366
Test Specificity Cross reacts with Human. Expected from sequence similarity: Mouse and Dog.
Target Species Human, Mouse, Rat, Rabbit
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.