Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UBXD2 Antibody, Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB145343 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB145343 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB145343 Supplier Novus Biologicals Supplier No. NBP305991AF488
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

UBXD2 Polyclonal specifically detects UBXD2 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen UBXD2
Applications Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 488
Dilution Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias erasin, FLJ23318, KIAA0242KIAA2042, UBX domain containing 2, UBX domain protein 4, UBX domain-containing 1, UBX domain-containing 2, UBX domain-containing protein 2, UBX domain-containing protein 4, UBXD2, UBXDC1
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human UBXD2 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23190
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.