Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UBXD5 Antibody, Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. NB144560 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB144560 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB144560 Supplier Novus Biologicals Supplier No. NBP305951AF647
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

UBXD5 Polyclonal specifically detects UBXD5 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen UBXD5
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 647
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias COA-1, Colorectal tumor-associated antigen COA-1, colorectal tumor-associated antigen-1, SOCDKFZp686F04228, SOCI, socius, UBX domain containing 5, UBX domain protein 11, UBX domain-containing protein 11, UBX domain-containing protein 5, UBXD5
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human UBXD5 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 91544
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.