Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UBXN2A Antibody, Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB320306 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB320306 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB320306 Supplier Novus Biologicals Supplier No. NBP298564APC
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

UBXN2A Polyclonal antibody specifically detects UBXN2A in Human samples. It is validated for Western Blot

Specifications

Antigen UBXN2A
Applications Western Blot
Classification Polyclonal
Conjugate APC
Dilution Western Blot
Formulation PBS
Gene Alias MGC138202, UBX domain containing 4, UBX domain protein 2A, UBX domain-containing protein 2A, UBX domain-containing protein 4, UBXD4
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human UBXN2A fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 165324
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.