Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UBXN2B Antibody (OTI1F7), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB280722 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB280722 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB280722 Supplier Novus Biologicals Supplier No. NBP274769MFV450

Mouse Monoclonal Antibody

UBXN2B Monoclonal antibody specifically detects UBXN2B in Human, Mouse, Rat samples. It is validated for Flow Cytometry

Specifications

Antigen UBXN2B
Applications Flow Cytometry
Classification Monoclonal
Clone OTI1F7
Conjugate mFluor Violet 450 SE
Dilution Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias UBX domain protein 2B
Host Species Mouse
Immunogen Full length human recombinant protein of human UBXN2B (NP_001071087) produced in HEK293T cell.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 137886
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.