Learn More
Description
Specifications
Specifications
| Antigen | UBXN8 |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 0.4 ug/ml, Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 |
| Formulation | PBS, pH 7.2, containing 40% glycerol with 0.02% sodium azide |
| Gene Alias | D8S2298EUBXD6, Rep-8 protein, REP8UBX domain-containing protein 6, Reproduction 8 protein, Reproduction/chromosome 8, UBX domain containing 6, UBX domain protein 8, UBX domain-containing protein 8 |
| Gene Symbols | UBXN8 |
| Host Species | Rabbit |
| Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids: GDEGTSQTSFETSNREAAKSQNLPKPLTEFPSPAEQPTCKEIPDLPEEPSQTAEEVVTVALRCPSGNVLRRRFLKSYSSQVLFDWMTRIGYHISLYSLSTSFPRRPLAVEGGQSLEDIGITVDTVLILEEKEQTN |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
