Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ UCB 35625
GREENER_CHOICE

Catalog No. 27571
Change view
Click to view available options
Quantity:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
27-571 1 mg
27-571-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 27-571 Supplier Tocris Bioscience™ Supplier No. 2757/1
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent CCR1 and CCR3 antagonist

Potent chemokine CCR1 and CCR3 receptor antagonist. Inhibits MIP-1α-induced chemotaxis in CCR1 transfectants and eotaxin-induced chemotaxis in CCR3 transfectants (IC50 values are 9.57 and 93.8 nM respectively). Antagonizes CCR3-mediated entry of HIV-1 isolate 89.6 into NP-2 cells (IC50 = 57 nM).

Isomer also available.

Chemical Identifiers

CAS 202796-42-7
Molecular Formula C30H37Cl2IN2O2

Specifications

Quantity 1 mg
Formula Weight 655.44
Purity >98%
Molecular Weight (g/mol) 655.44
Chemical Name or Material UCB 35625
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.