Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UCH-L1 Mouse anti-Human, Clone: 972106, Novus Biologicals™
SDP

Catalog No. MAB60071100 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB60071100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB60071100 Supplier Novus Biologicals Supplier No. MAB60071100UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

UCH-L1 Monoclonal specifically detects UCH-L1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), Simple Western, Western Blot.

Specifications

Antigen UCH-L1
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Simple Western, Western Blot
Classification Monoclonal
Clone 972106
Conjugate Unconjugated
Dilution Western Blot, Simple Western, Immunohistochemistry
Formulation PBS with No Preservative
Gene Alias EC 3.4.19.12, Neuron cytoplasmic protein 9.5, PARK5, PGP 9.5, PGP9.5, PGP95, ubiquitin carboxyl-terminal esterase L1 (ubiquitin thiolesterase), ubiquitin carboxyl-terminal hydrolase isozyme L1, ubiquitin C-terminal hydrolase, Ubiquitin thioesterase L1, UCH-L1
Gene Symbols UCHL1
Host Species Mouse
Immunogen E. coli-derived recombinant human UCH-L1/PGP9.5
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7345
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.