Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UCH-L1/PGP9.5 Antibody, Novus Biologicals™
SDP

Catalog No. NB300676SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB300676SS 0.025 mL
NB300676 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB300676SS Supplier Novus Biologicals Supplier No. NB300676SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 2 publications

UCH-L1/PGP9.5 Polyclonal specifically detects UCH-L1/PGP9.5 in Human, Mouse samples. It is validated for Flow Cytometry, Gene Knock-Out, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Simple Western, Western Blot.

Specifications

Antigen UCH-L1/PGP9.5
Applications Flow Cytometry, Gene Knock-Out, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Simple Western, Western Blot
Classification Polyclonal
Concentration 1.0 mg/ml
Conjugate Unconjugated
Dilution Western Blot 2 ug/mL, Simple Western 1:2000, Flow Cytometry 2-5 ug/million cells/ml, Immunohistochemistry 2.5 - 5.0 ug/mL, Immunocytochemistry/Immunofluorescence 1:100 - 1:200, Immunohistochemistry-Paraffin 2.5 - 5.0 ug/mL, Immunohistochemistry-Frozen reported in scientific literature (PMID 25041530), Knockout Validated
Formulation Tris-Glycine, 0.15 M NaCl with 0.05% Sodium Azide
Gene Accession No. P09936
Gene Alias EC 3.4.19.12, EC 6.-, Neuron cytoplasmic protein 9.5, PARK5, PGP 9.5, PGP9.5, PGP9.5Uch-L1, PGP95, ubiquitin carboxyl-terminal esterase L1 (ubiquitin thiolesterase), ubiquitin carboxyl-terminal hydrolase isozyme L1, ubiquitin C-terminal hydrolase, Ubiquitin thioesterase L1, UCHL1, UCH-L1
Gene Symbols UCHL1
Host Species Rabbit
Immunogen A synthetic peptide made to a C-terminal portion of the human UCHL1 protein (between residues 200-220). [UniProt# P09936]
Molecular Weight of Antigen 25 kDa
Purification Method Affinity Purified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Alzheimers Research, Cellular Markers, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Neurotransmission
Primary or Secondary Primary
Gene ID (Entrez) 7345
Test Specificity Human and mouse. Predicted to react with monkey, rat, pig, and horse based on 100% sequence homology.
Target Species Human, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.