Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UCH-L1/PGP9.5 Antibody (BH7), Novus Biologicals™
SDP

Catalog No. NB11058869 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB11058869 0.1 mL
NB017972 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB11058869 Supplier Novus Biologicals Supplier No. NB11058869
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

UCH-L1/PGP9.5 Monoclonal specifically detects UCH-L1/PGP9.5 in Human, Mouse, Rat, Porcine, Bovine, Equine samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry Free-Floating.

Specifications

Antigen UCH-L1/PGP9.5
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Free Floating)
Classification Monoclonal
Clone BH7
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1:20000, Simple Western 1:20000, Immunohistochemistry 1:2000, Immunocytochemistry/Immunofluorescence 1:2000, Immunohistochemistry Free-Floating 1:2000
Formulation 50% PBS, 50% glycerol with 5mM Sodium Azide
Gene Accession No. P09936
Gene Alias EC 3.4.19.12, EC 6.-, Neuron cytoplasmic protein 9.5, PARK5, PGP 9.5, PGP9.5, PGP9.5Uch-L1, PGP95, ubiquitin carboxyl-terminal esterase L1 (ubiquitin thiolesterase), ubiquitin carboxyl-terminal hydrolase isozyme L1, ubiquitin C-terminal hydrolase, Ubiquitin thioesterase L1, UCHL1, UCH-L1
Gene Symbols UCHL1
Host Species Mouse
Immunogen Recombinant full length human UCHL1 purified from E. coli. [Swiss-Prot# P09936]
Molecular Weight of Antigen 24 kDa
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuronal Cell Markers, Neuroscience, Neurotransmission
Primary or Secondary Primary
Gene ID (Entrez) 7345
Target Species Human, Mouse, Rat, Pig, Bovine, Equine
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Ascites
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.