Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
UCH-L1/PGP9.5 Antibody (BH7), Novus Biologicals™

Mouse Monoclonal Antibody
$206.50 - $499.50
Specifications
Antigen | UCH-L1/PGP9.5 |
---|---|
Clone | BH7 |
Concentration | 1 mg/ml |
Dilution | Western Blot 1:20000, Simple Western 1:20000, Immunohistochemistry 1:2000, Immunocytochemistry/Immunofluorescence 1:2000, Immunohistochemistry Free-Floating 1:2000 |
Applications | Immunohistochemistry (Free Floating), Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB017972
![]() |
Novus Biologicals
NB110588690.025ML |
0.025 mL |
Each for $206.50
|
|
|||||
NB11058869
![]() |
Novus Biologicals
NB11058869 |
0.1 mL |
Each for $499.50
|
|
|||||
Description
UCH-L1/PGP9.5 Monoclonal specifically detects UCH-L1/PGP9.5 in Human, Mouse, Rat, Porcine, Bovine, Equine samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry Free-Floating.Specifications
UCH-L1/PGP9.5 | |
1 mg/ml | |
Immunohistochemistry (Free Floating), Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
Mouse | |
Neuronal Cell Markers, Neuroscience, Neurotransmission | |
P09936 | |
7345 | |
Recombinant full length human UCHL1 purified from E. coli. [Swiss-Prot# P09936] | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
BH7 | |
Western Blot 1:20000, Simple Western 1:20000, Immunohistochemistry 1:2000, Immunocytochemistry/Immunofluorescence 1:2000, Immunohistochemistry Free-Floating 1:2000 | |
Monoclonal | |
Ascites | |
RUO | |
Human, Rat, Bovine, Mouse, Equine, Pig | |
EC 3.4.19.12, EC 6.-, Neuron cytoplasmic protein 9.5, PARK5, PGP 9.5, PGP9.5, PGP9.5Uch-L1, PGP95, ubiquitin carboxyl-terminal esterase L1 (ubiquitin thiolesterase), ubiquitin carboxyl-terminal hydrolase isozyme L1, ubiquitin C-terminal hydrolase, Ubiquitin thioesterase L1, UCHL1, UCH-L1 | |
UCHL1 | |
IgG1 | |
Affinity purified | |
24 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title