Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UCH-L1/PGP9.5 Antibody, Novus Biologicals™
SDP

Catalog No. NB11058872T Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB11058872T 0.025 mL
NB11058872 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB11058872T Supplier Novus Biologicals Supplier No. NB11058872SS
Add to Cart
Add to Cart

Chicken Polyclonal Antibody

UCH-L1/PGP9.5 Polyclonal specifically detects UCH-L1/PGP9.5 in Human, Mouse, Rat, Porcine, Bovine, Equine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.

Specifications

Antigen UCH-L1/PGP9.5
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:10000, Immunohistochemistry 1:1000, Immunocytochemistry/Immunofluorescence 1:1000, Immunohistochemistry-Frozen
Formulation Supplied as an aliquot of IgY preparation with 5mM Sodium Azide
Gene Accession No. P09936
Gene Alias EC 3.4.19.12, EC 6.-, Neuron cytoplasmic protein 9.5, PARK5, PGP 9.5, PGP9.5, PGP9.5Uch-L1, PGP95, ubiquitin carboxyl-terminal esterase L1 (ubiquitin thiolesterase), ubiquitin carboxyl-terminal hydrolase isozyme L1, ubiquitin C-terminal hydrolase, Ubiquitin thioesterase L1, UCHL1, UCH-L1
Gene Symbols UCHL1
Host Species Chicken
Immunogen Recombinant full length human UCHL1 purified from E. coli. [UniProt# P09936]
Molecular Weight of Antigen 24 kDa
Purification Method Ammonium sulfate precipitation
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Alzheimers Research, Cellular Markers, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Neurotransmission
Primary or Secondary Primary
Gene ID (Entrez) 7345
Target Species Human, Mouse, Rat, Pig, Bovine, Equine
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Precipitation
Isotype IgY
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.