Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UCH-L3 Antibody, FITC, Novus Biologicals™
SDP

Catalog No. NB082631 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB082631 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB082631 Supplier Novus Biologicals Supplier No. NBP298421F
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

UCH-L3 Polyclonal specifically detects UCH-L3 in Rat samples. It is validated for ELISA.

Specifications

Antigen UCH-L3
Applications ELISA
Classification Polyclonal
Conjugate FITC
Formulation PBS
Gene Alias EC 3.4.19.12, ubiquitin carboxyl-terminal esterase L3 (ubiquitin thiolesterase), ubiquitin carboxyl-terminal hydrolase isozyme L3, Ubiquitin thioesterase L3, ubiquitin thiolesterase, UCH-L3
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant UCH-L3 (Q91Y78; Glu 2-Ala 230).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7347
Target Species Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.