Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UCH-L3 Rabbit anti-Mouse, Clone: 204, Novus Biologicals™
SDP

Catalog No. NB025107 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB025107 100 μg
NB025108 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB025107 Supplier Novus Biologicals Supplier No. NBP290680100UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

UCH-L3 Monoclonal specifically detects UCH-L3 in Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen UCH-L3
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 204
Conjugate Unconjugated
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin 1:100-1:500
Formulation 0.2 um filtered solution in PBS
Gene Alias EC 3.4.19.12, ubiquitin carboxyl-terminal esterase L3 (ubiquitin thiolesterase), ubiquitin carboxyl-terminal hydrolase isozyme L3, Ubiquitin thioesterase L3, ubiquitin thiolesterase, UCH-L3
Host Species Rabbit
Immunogen Recombinant Mouse UCH-L3 protein
Purification Method Protein A purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7347
Target Species Mouse
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.