Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UCN2 Rabbit anti-Human, Polyclonal, Proteintech
SDP

Catalog No. p-7204416 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-172-7786 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-172-7786 Supplier Proteintech Group Inc Supplier No. 121981AP150UL
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Two major regulatory peptides were originally isolated from fish urophysial extracts, Urotensin I and II. In both frog and human, the Urotensin II sequence is located at the carboxy-terminal position of the precursor. Human Urotensin II is composed of only 11 amino acid residues, while fish and frog Urotensin II possess 12 and 13 amino acid residues, respectively. The cyclic region of Urotensin II, which is responsible for the biological activity of the peptide, has been fully conserved from fish to human. However, several substitutions have occurred in the amino-terminal region of the molecule. A human G protein-coupled receptor, GPR14, is the Urotensin II receptor. Human Urotensin II is found within both vascular and cardiac tissue, including coronary atheroma, and effectively constricts isolated arteries from nonhuman primates. Urotensin II may act as an autocrine and/or paracrine hormone rather than as a circulating hormone, by playing an important role in the development of ventricular hypertrophy induced by chronic hypoxia.

Specifications

Antigen UCN2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.37 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene UCN2
Gene Accession No. Q96RP3
Gene Alias SRP, Stresscopin related peptide, UCN II, UCN2, UCNI, UR, urocortin 2, Urocortin II, Urocortin related peptide, URP
Gene Symbols UCN2
Host Species Rabbit
Immunogen UCN2 Fusion Protein Ag2836
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 90226
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.