Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UCP1/3 Antibody (SR1570), Novus Biologicals™
SDP

Catalog No. NB258796 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB258796 100 μg
NB259290 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB258796 Supplier Novus Biologicals Supplier No. NBP322657100UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

UCP1/3 Monoclonal antibody specifically detects UCP1/3 in Mouse, Rat samples. It is validated for Western Blot, Immunoprecipitation

Specifications

Antigen UCP1/3
Applications Western Blot, Immunoprecipitation
Classification Monoclonal
Clone SR1570
Conjugate Unconjugated
Dilution Western Blot 1:500-1:1000, Immunoprecipitation 1:50
Formulation PBS, pH 7.4, 150mM NaCl, 50% glycerol.
Host Species Rabbit
Immunogen A synthesized peptide derived from human UCP1/3 (Uniprot #: P12242, P56501)
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse, Rat
Content And Storage Store at -20° C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.