Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UCP1 Antibody, Novus Biologicals™
SDP

Catalog No. NB434181 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB434181 25 μg
NB1002828 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB434181 Supplier Novus Biologicals Supplier No. NB100282825UG
Add to Cart
Add to Cart

Goat Polyclonal Antibody has been used in 3 publications

UCP1 Polyclonal specifically detects UCP1 in Human, Mouse, Rat samples. It is validated for IF/IHC, Immunohistochemistry, Peptide ELISA, Western Blot.

Specifications

Antigen UCP1
Applications Immunohistochemistry, Peptide ELISA, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.5 - 1 μg/mL, Peptide ELISA Detection limit 1:128000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_068605.1
Gene Alias SLC25A7mitochondrial brown fat uncoupling protein 1, Solute carrier family 25 member 7, thermogenin, UCP, UCP 1, uncoupling protein 1 (mitochondrial, proton carrier)
Gene Symbols UCP1
Host Species Goat
Immunogen Peptide with sequence C-EQLKRELSKSRQ corresponding to C-Terminus according to NP_068605.1.
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Cancer, Cellular Signaling, metabolism
Primary or Secondary Primary
Gene ID (Entrez) 7350
Test Specificity Expected from sequence similarity: Rat and Dog.
Target Species Human, Rat, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.