Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UCP1 Antibody (Vab12 P4B12*A12), Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NP20343A70 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP20343A70 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP20343A70 Supplier Novus Biologicals Supplier No. NBP250343AF750
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

UCP1 Monoclonal specifically detects UCP1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.

Specifications

Antigen UCP1
Applications Western Blot, ELISA, Immunohistochemistry
Classification Monoclonal
Clone Vab12 P4B12*A12
Conjugate Alexa Fluor 750
Dilution Western Blot, ELISA, Immunohistochemistry
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias SLC25A7mitochondrial brown fat uncoupling protein 1, Solute carrier family 25 member 7, thermogenin, UCP, UCP 1, uncoupling protein 1 (mitochondrial, proton carrier)
Gene Symbols UCP1
Host Species Mouse
Immunogen Ovalbumin-conjugated synthetic peptide - (10 amino acids) derived from the middle polypeptide region of human UCP1. The exact sequence is proprietary.
Purification Method Protein A purified
Quantity 0.1 mL
Research Discipline Cancer, Cellular Signaling, metabolism
Primary or Secondary Primary
Gene ID (Entrez) 7350
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.