Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UCRC Monoclonal Antibody (2B5)
GREENER_CHOICE

Catalog No. PIMA521779
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA521779 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA521779 Supplier Invitrogen™ Supplier No. MA521779
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: MAAATLTSKL YSLLFRRTST FALTIIVGVM FFERAFDQGA DAIYDHINEG KLWKHIKHKY ENK.

UCRC is a subunit of mitochondrial complex III (ubiquinol-cytochrome c reductase; EC 1.10.2.2), which forms the middle segment of the respiratory chain of the inner mitochondrial membrane. UCRC is a subunit of mitochondrial complex III (ubiquinol-cytochrome c reductase; EC 1.10.2.2), which forms the middle segment of the respiratory chain of the inner mitochondrial membrane (Schagger et al., 1995 ).
TRUSTED_SUSTAINABILITY

Specifications

Antigen UCRC
Applications ELISA
Classification Monoclonal
Clone 2B5
Concentration 0.2-1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene UQCR10
Gene Accession No. Q9UDW1
Gene Alias 1110020P15Rik; AA960494; Complex III subunit 9; complex III subunit X; Cytochrome b-c1 complex subunit 9; cytochrome c1 non-heme 7 kDa protein; cytochrome C1, nonheme 7kDa protein; HSPC051; HSPC119; HSPC151; QCR9; ubiquinol-cytochrome c reductase complex (7.2 kD); ubiquinol-cytochrome c reductase complex 7.2 kDa protein; ubiquinol-cytochrome c reductase complex 7.2kDa protein; ubiquinol-cytochrome c reductase complex protein; ubiquinol-cytochrome c reductase, complex III subunit X; ubiquinol-cytochrome c reductase, complex III subunit X, 7.2kDa; UCCR7.2; Ucrc; Unknown (protein for MGC:127828); UQCR10
Gene Symbols UQCR10
Host Species Mouse
Immunogen UQCR10 (AAH05402, 1 a.a. ~ 63 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29796
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.