Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UDP glucose dehydrogenase Antibody (2G11), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NB161780 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB161780 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB161780 Supplier Novus Biologicals Supplier No. NBP259461AF350
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

UDP glucose dehydrogenase Monoclonal antibody specifically detects UDP glucose dehydrogenase in Human samples. It is validated for Western Blot, ELISA, Immunofluorescence

Specifications

Antigen UDP glucose dehydrogenase
Applications Western Blot, ELISA, Immunofluorescence
Classification Monoclonal
Clone 2G11
Conjugate Alexa Fluor 350
Formulation 50mM Sodium Borate
Gene Alias EC 1.1.1.22, GDH, UDPGDHUDP-glucose dehydrogenase, UDP-Glc dehydrogenase, UDP-GlcDH, UDP-glucose 6-dehydrogenase, UGD, uridine diphospho-glucose dehydrogenase
Host Species Mouse
Immunogen Recombinant human UDP glucose dehydrogenase (1-494aa) purified from E. coli
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 7358
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.