Learn More
Thermo Scientific™ μDrop™ and μDrop Duo Plates
Catalog No. 501903760
Encompass
Click to view available options
Product Type:
μDrop Plate
Catalog No. 50-190-3760
Supplier: Thermo Scientific™ N12391
Only null left
Add to Cart
Add to Cart
Thermo Scientific™ μDrop and μDrop Duo plates are used for low volume (2-10 μl) photometric DNA, RNA and protein quantitation and purity analysis. The μDrop plates are compatible with Thermo Scientific Varioskan LUX and Multiskan SkyHigh microplate readers. The ready-made protocols in SkanIt software or Multiskan SkyHigh user interface make quantification fast and easy.
Thermo Scientific™ μDrop™ and μDrop™ Duo Plates are ideal tools for quick and easy photometric low-volume (2–10 μl) concentration measurements of DNA, RNA, protein samples, or spectral scanning. The low volume area is designed to be used with 8-channel pipette for fast and accurate multiple sample pipetting. The plates also contain a separate area for cuvette measurement. μDrop Plates are compatible with Thermo Scientific Varioskan™ LUX, Multiskan™ Sky and Multiskan SkyHigh microplate readers. The intuitive Thermo Scientific SkanIt™ software and Multiskan SkyHigh user interface offer ready-made measurement protocols for quick sample measurement and analysis.
  • dsDNA detection range from a few micrograms to a few milligrams per milliliter
  • Sixteen sample positions for quick and easy measurement of sample volumes down to 2μL
  • Compatible with an eight-channel pipette for easy sample addition
  • Quick and easy to wipe off the samples in serial measurements
  • Ready-made Thermo Scientific SkanIt Software sessions for nucleic acid analysis
  • Measurement slot for a rectangular cuvette
  • Compatible with Multiskan Sky, Multiskan GO, Varioskan LUX and Varioskan Flash
TRUSTED_SUSTAINABILITY
Specifications
Product Type μDrop Plate
Plate Type μDrop Duo Plate
Capacity (Metric) 2 to 10μL (low-volume function)
No. of Positions 16
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.