Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UEV2 Antibody, Novus Biologicals™
SDP

Catalog No. NB10056360 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10056360 0.1 mg
NB10056360T 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10056360 Supplier Novus Biologicals Supplier No. NB10056360
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

UEV2 Polyclonal specifically detects UEV2 in Human samples. It is validated for Western Blot.

Specifications

Antigen UEV2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 2 ug/ml
Formulation PBS containing 0.2% gelatin with 0.05% Sodium Azide
Gene Accession No. NP_003341
Gene Alias 1 alpha, 25-dihydroxyvitamin D3-inducible, DDVit-1, DDVIT1, EDAF-1, EDPF1, EDPF-1DDVit 1, enterocyte differentiation promoting factor, Enterocyte differentiation-associated factor 1, enterocyte differentiation-associated factor EDAF-1, Enterocyte differentiation-promoting factor 1, methyl methanesulfonate sensitive 2, S. cerevisiae, homolog of, MMS2MMS2 homolog, ubiquitin-conjugating enzyme E2 variant 2, UEV2, UEV-2, Vitamin D3-inducible protein
Gene Symbols UBE2V2
Host Species Rabbit
Immunogen This antibody was generated by immunizing rabbit with a mixture of synthetic peptides containing amino acids 12-28 of human UBE2V2.
Purification Method Protein G purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 7336
Test Specificity UBE2V2
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.