Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UFL1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB167174 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB167174 20 μg
NB167173 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB167174 Supplier Novus Biologicals Supplier No. NBP31603920UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

UFL1 Polyclonal antibody specifically detects UFL1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunofluorescence

Specifications

Antigen UFL1
Applications Western Blot, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500 - 1:2000, Immunocytochemistry/Immunofluorescence 1:50 - 1:200
Formulation PBS, 50% glycerol, pH7.3
Gene Alias E3 UFM1-protein ligase 1, EC 6.3.2.-, KIAA0776, LZAP-binding protein, NLBPnovel LZAP-binding protein, RP3-393D12.1, UFL1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 601-794 of human UFL1 (NP_056138.1). AAITSEIRKKILSKLSEETKVALTKLHNSLNEKSIEDFISCLDSAAEACDIMVKRGDKKRERQILFQHRQALAEQLKVTEDPALILHLTSVLLFQFSTHSMLHAPGRCVPQIIAFLNSKIPEDQHALLVKYQGLVVKQLVSQSKKTGQGDYPLNNELDKEQEDVASTTRKELQELSSSIKDLVLKSRKSSVTEE
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23376
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.