Learn More
Stoelting™ Ugo Basile Multi-Maze
SDP
Stoelting™ Ugo Basile Multi-Maze for Mice is a modular system providing full automation of Y-maze, T-maze and 8-arm Radial Arm Maze.
$6670.00 - $7120.00
Specifications
For Use With (Application) Neuroscience
View More Specs
Products 6
Catalog Number Mfr. No. Animal Model Price Quantity & Availability  
Catalog Number Mfr. No. Animal Model Price Quantity & Availability  
10-001-098
SDP
View Documents
Stoelting™
675031
Mouse Central arena
Each for $7,120.00
Only null left
Add to Cart
10-001-099
SDP
View Documents
Stoelting™
675032
Mouse Standard arm with automated door
Each for $6,910.00
Only null left
Add to Cart
10-001-100
SDP
View Documents
Stoelting™
675033
Mouse Start compartment w/automated door
Each for $6,690.00
Only null left
Add to Cart
10-001-101
SDP
View Documents
Stoelting™
675061
Rat Central arena
Each for $6,910.00
Only null left
Add to Cart
10-001-102
SDP
View Documents
Stoelting™
675062
Rat Standard arm with automated door
Each for $6,960.00
Only null left
Add to Cart
10-001-103
SDP
View Documents
Stoelting™
675063
Rat Start compartment w/automated door
Each for $6,670.00
Only null left
Add to Cart
Description
Description
Allows for conducting fully automated memory experiments including:
  • Assessment of spatial memory
  • Basic working memory
  • Discriminating working from reference memory
  • Evaluating impairments in working memory
Includes a central electronic unit and up to eight modular units that can be configured as:
  • Y-maze
  • T-maze
  • 8-arm Radial Arm Maze
Proprietary sliding doors retract into the maze floor smoothly and silently without disrupting the animal during testing. This sliding mechanism combined with the non-reflective, high contrast maze material provides optimal results with ANY-maze Video Tracking Software (sold separately).
Specifications
Specifications
Neuroscience
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.