Learn More
Stoelting™ Ugo Basile Sociability Apparatus
SDP
Stoelting™ Sociability Apparatus is a 3-chambered social test used to study social behaviour in mice. Useful for research on autism, parental behavior, sociability and social memory (social novelty).
$5600.00 - $8180.00
Specifications
For Use With (Application) Neuroscience
View More Specs
Products 5
Catalog Number Mfr. No. Animal Model Price Quantity & Availability  
Catalog Number Mfr. No. Animal Model Price Quantity & Availability  
10-001-061
SDP
View Documents
Stoelting™
60448
Rat Opaque with gray enclosures
Each for $8,180.00
Only null left
Add to Cart
10-001-062
SDP
View Documents
Stoelting™
60449
Rat With white enclosures
Each for $8,180.00
Only null left
Add to Cart
10-001-063
SDP
View Documents
Stoelting™
60450
Mouse With 2 stranger enclosures
Each for $5,820.00
Only null left
Add to Cart
10-001-065
SDP
View Documents
Stoelting™
60456
Mouse With white enclosures
Each for $5,820.00
Only null left
Add to Cart
10-001-067
SDP
View Documents
Stoelting™
60458
Mouse Opaque with gray enclosures
Each for $5,600.00
Only null left
Add to Cart
Description
Description
Ideal for the investigation of autism-related behaviors:
  • Social approach
  • Resistance to reverse a learned pattern of behavior
Shortly after a period of habituation a mouse's sociability is determined by evaluating the amount of time the test mouse spends approaching a wire cage containing an unfamiliar stranger mouse.
In the second phase of testing a second unfamiliar mouse is introduced into the Sociability Apparatus. The preference for social novelty is then determined by measuring the amount of time the test mouse spends approaching the chamber containing a new, unfamiliar stranger mouse (also restricted to a small wire cage), as compared to time spent in the chamber containing the now-familiar mouse. Resistance to change is measured by two reversal learning tasks.
Testing in the Sociability Apparatus can be completely automated by the ANY-maze software system.
Specifications
Specifications
Neuroscience
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.