Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UGP2 Mouse anti-Human, Mouse, Rat, Clone: 7H1, Novus Biologicals™
SDP

Catalog No. NBP275695 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP275695 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP275695 Supplier Novus Biologicals Supplier No. NBP275695

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse Monoclonal Antibody

UGP2 Monoclonal specifically detects UGP2 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen UGP2
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 7H1
Concentration 2 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10 - 1:500, Immunohistochemistry-Paraffin
Formulation PBS (pH7.4), 0.2% BSA, 50% Glycerol with 0.05% Sodium Azide
Gene Alias EC 2.7.7.9, pHC379, UDPG, UDP-glucose diphosphorylase, UDP-glucose pyrophosphorylase, UDP-glucose pyrophosphorylase 2, UDPGP, UDPGP2, UGP1, UGPase, UGPase 2, UGPP2, uridyl diphosphate glucose pyrophosphorylase 2, UTP-glucose-1-phosphate uridyltransferase, UTP--glucose-1-phosphate uridylyltransferase, UTP--glucose-1-phosphate uridylyltransferase 2
Gene Symbols UGP2
Host Species Mouse
Immunogen Synthetic peptide within Human UGP2 aa 61-110/ 508. (SwissProt: Q16851 Human; SwissProt: Q91ZJ5 Mouse)
Molecular Weight of Antigen 57 kDa
Purification Method Protein G purified
Quantity 100 μL
Research Discipline Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 7360.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.