Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UGT1A6 Antibody, Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NB049113 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB049113 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB049113 Supplier Novus Biologicals Supplier No. NBP297897AF350
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

UGT1A6 Polyclonal specifically detects UGT1A6 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen UGT1A6
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 350
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias GNT1UDPGT 1-6, HLUGP, HLUGP1, MGC29860, Phenol-metabolizing UDP-glucuronosyltransferase, UDP glucuronosyltransferase 1 family, polypeptide A6, UDP glycosyltransferase 1 family, polypeptide A6, UDP-glucuronosyltransferase 1 family polypeptide A6s, UDP-glucuronosyltransferase 1-6, UDP-glucuronosyltransferase 1A6, UDP-glucuronosyltransferase 1-F, UDPGT, UGT1, UGT1*6, UGT1.6, UGT1-06, UGT1A6S, UGT-1F, UGT1FEC 2.4.1.17
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human UGT1A6 fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 54578
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.