Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UGT2B4 Antibody, Novus Biologicals™
SDP

Catalog No. NBP254718 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP254718 100 μL
NB404097 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP254718 Supplier Novus Biologicals Supplier No. NBP254718
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

UGT2B4 Polyclonal specifically detects UGT2B4 in Human, Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen UGT2B4
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry-Paraffin 1:500 - 1:1000
Formulation PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide
Gene Alias family 2, beta-4, UDP glucuronosyltransferase 2 family, polypeptide B4, UDP glycosyltransferase 2 family, polypeptide B4
Gene Symbols UGT2B4
Host Species Rabbit
Immunogen This antibody was developed against a Recombinant Protein corresponding to amino acids:FVGGLHCKPAKPLPKEMEEFVQSSG
Purification Method Immunogen affinity purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7363
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.