Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UGT3A1 Antibody, Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB353003 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB353003 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB353003 Supplier Novus Biologicals Supplier No. NBP306222APC
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

UGT3A1 Polyclonal antibody specifically detects UGT3A1 in Human samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen UGT3A1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate APC
Dilution Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias EC 2.4.1, EC 2.4.1.17, FLJ26528, FLJ34658, UDP glycosyltransferase 3 family, polypeptide A1, UDP-glucuronosyltransferase 3A1, UDPGT 3A1
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human UGT3A1 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 133688
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.