Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UHRF1 Antibody (1A1B1) - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP261838 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP2-61838 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP2-61838 Supplier Novus Biologicals Supplier No. NBP261838
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

UHRF1 Monoclonal specifically detects UHRF1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen UHRF1
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 1A1B1
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000
Formulation PBS with 0.05% Sodium Azide
Gene Alias E3 ubiquitin-protein ligase UHRF1, EC 6.3.2, EC 6.3.2.-, FLJ21925, hNP95, huNp95, ICBP90NP95, Inverted CCAAT box-binding protein of 90 kDa, Np95, Nuclear protein 95, Nuclear zinc finger protein Np95, RING finger protein 106, RNF106MGC138707, Transcription factor ICBP90, Ubiquitin-like PHD and RING finger domain-containing protein 1, ubiquitin-like with PHD and ring finger domains 1, ubiquitin-like, containing PHD and RING finger domains, 1, Ubiquitin-like-containing PHD and RING finger domains protein 1
Gene Symbols UHRF1
Host Species Mouse
Immunogen Purified recombinant fragment of human UHRF1 (AA: 616-755) expressed in E. Coli.
Molecular Weight of Antigen 91.3 kDa
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline DNA Repair, Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 29128
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.