Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
UHRF1 Antibody (1A1B1) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$560.50
Specifications
Antigen | UHRF1 |
---|---|
Clone | 1A1B1 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000 |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP2-61838
![]() |
Novus Biologicals
NBP261838 |
0.1 mL |
Each of 1 for $560.50
|
|
|||||
Description
UHRF1 Monoclonal specifically detects UHRF1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
UHRF1 | |
1 mg/mL | |
Monoclonal | |
Purified | |
DNA Repair, Zinc Finger | |
29128 | |
Purified recombinant fragment of human UHRF1 (AA: 616-755) expressed in E. Coli. | |
Primary | |
91.3 kDa |
1A1B1 | |
Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000 | |
Unconjugated | |
Mouse | |
E3 ubiquitin-protein ligase UHRF1, EC 6.3.2, EC 6.3.2.-, FLJ21925, hNP95, huNp95, ICBP90NP95, Inverted CCAAT box-binding protein of 90 kDa, Np95, Nuclear protein 95, Nuclear zinc finger protein Np95, RING finger protein 106, RNF106MGC138707, Transcription factor ICBP90, Ubiquitin-like PHD and RING finger domain-containing protein 1, ubiquitin-like with PHD and ring finger domains 1, ubiquitin-like, containing PHD and RING finger domains, 1, Ubiquitin-like-containing PHD and RING finger domains protein 1 | |
UHRF1 | |
IgG2b | |
Protein G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title