Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UHRF1BP1L Antibody, Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB184123 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB184123 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB184123 Supplier Novus Biologicals Supplier No. NBP282047AF488
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

UHRF1BP1L Polyclonal antibody specifically detects UHRF1BP1L in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen UHRF1BP1L
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 488
Formulation 50mM Sodium Borate
Gene Alias KIAA0701UHRF1 (ICBP90) binding protein 1-like, UHRF1 binding protein 1-like, UHRF1-binding protein 1-like
Host Species Rabbit
Immunogen UHRF1BP1L antibody was raised against a 17 amino acid synthetic peptide near the center of human UHRF1BP1L. The immunogen is located within amino acids 980 - 1030 of UHRF1BP1L. Amino Acid Squence: EIGSETSDKKDSFYTDS
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 23074
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.