Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
UIP1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP155442
Description
UIP1 Polyclonal specifically detects UIP1 in Human samples. It is validated for Western Blot.Specifications
| UIP1 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| HAUS7 | |
| Synthetic peptides corresponding to UCHL5IP(UCHL5 interacting protein) The peptide sequence was selected from the middle region of UCHL5IP. Peptide sequence LLDTIRSLTIGCSSCSSLMEHFEDTREKNEALLGELFSSPHLQMLLNPEC. | |
| Affinity purified | |
| RUO | |
| 55559 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| 26S proteasome-associated UCH37-interacting protein 1, expressed-Xq28STS protein, HAUS augmin-like complex subunit 7, HAUS augmin-like complex, subunit 7, UCH37 interacting protein 1, UCHL5 interacting protein, UCHL5-interacting protein, UCHL5IPHSXQ28ORF, UIP126S proteasome-associated UCH interacting protein 1, X-linked protein STS1769 | |
| Rabbit | |
| 47 kDa | |
| 100 μL | |
| Primary | |
| This product is specific to Subunit or Isoform: 7. | |
| Human, Mouse, Pig, Bovine, Canine, Equine, Rabbit | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction