Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
UIP1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
| Antigen | UIP1 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
UIP1 Polyclonal specifically detects UIP1 in Human samples. It is validated for Western Blot.Specifications
| UIP1 | |
| Polyclonal | |
| Rabbit | |
| 26S proteasome-associated UCH37-interacting protein 1, expressed-Xq28STS protein, HAUS augmin-like complex subunit 7, HAUS augmin-like complex, subunit 7, UCH37 interacting protein 1, UCHL5 interacting protein, UCHL5-interacting protein, UCHL5IPHSXQ28ORF, UIP126S proteasome-associated UCH interacting protein 1, X-linked protein STS1769 | |
| HAUS7 | |
| IgG | |
| This product is specific to Subunit or Isoform: 7. |
| Western Blot | |
| Unconjugated | |
| RUO | |
| 55559 | |
| Synthetic peptides corresponding to UCHL5IP(UCHL5 interacting protein) The peptide sequence was selected from the middle region of UCHL5IP. Peptide sequence LLDTIRSLTIGCSSCSSLMEHFEDTREKNEALLGELFSSPHLQMLLNPEC. | |
| Primary | |
| 47 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title