Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UIP1 Antibody (OTI1E8), FITC, Novus Biologicals™
SDP

Catalog No. NBP274782F Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP274782F 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP274782F Supplier Novus Biologicals Supplier No. NBP274782F
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

UIP1 Monoclonal specifically detects UIP1 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen UIP1
Applications Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI1E8
Conjugate FITC
Dilution Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation PBS with 0.05% Sodium Azide
Gene Alias 26S proteasome-associated UCH37-interacting protein 1, expressed-Xq28STS protein, HAUS augmin-like complex subunit 7, HAUS augmin-like complex, subunit 7, UCH37 interacting protein 1, UCHL5 interacting protein, UCHL5-interacting protein, UCHL5IPHSXQ28ORF, UIP126S proteasome-associated UCH interacting protein 1, X-linked protein STS1769
Gene Symbols HAUS7
Host Species Mouse
Immunogen Full length recombinant protein of human UIP1 (NP_059988) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 55559.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.