Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ UK 383367
GREENER_CHOICESDP

Catalog No. 418810 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
41-881-0 10 mg
41-881 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 41-881-0 Supplier Tocris Bioscience™ Supplier No. 4188/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent and selective BMP-1 (PCP) inhibitor

Potent and selective inhibitor of BMP-1 (procollagen C-proteinase; PCP) (IC50 = 44 nM). Selective for BMP-1 over MMPs 1, 2, 3, 9 and 14 (IC50 values are >10,000 nM for listed MMPs).

Chemical Identifiers

CAS 348622-88-8
Molecular Formula C15H24N4O4

Specifications

Quantity 10 mg
Formula Weight 324.38
Purity >99%
Molecular Weight (g/mol) 324.381
Chemical Name or Material UK 383367
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.