Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ULBP-1 Antibody (237104), Allophycocyanin/Cy7, Novus Biologicals™
SDP

Catalog No. NB313364 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB313364 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB313364 Supplier Novus Biologicals Supplier No. FAB2588APCCY7
Add to Cart
Add to Cart

Rat Monoclonal Antibody

ULBP-1 Monoclonal specifically detects ULBP-1 in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen ULBP-1
Applications Flow Cytometry
Classification Monoclonal
Clone 237104
Conjugate APC-Cyanine7
Dilution Flow Cytometry
Formulation PBS
Gene Alias ALCAN-beta, N2DL1, N2DL-1, NKG2D ligand 1, NKG2DL1, RAET1Ialcan-beta, Retinoic acid early transcript 1I, UL16 binding protein 1, UL16-binding protein 1
Host Species Rat
Immunogen BaF3 mouse pro-B cell line transfected with mouse ULBP-1/MULT-1
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 80329
Target Species Mouse
Content And Storage Store at 4C in the dark. Do not freeze.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.