Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ULBP-1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP220808 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP220808 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP220808 Supplier Novus Biologicals Supplier No. NBP220808
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ULBP-1 Polyclonal specifically detects ULBP-1 in Human samples. It is validated for Western Blot.

Specifications

Antigen ULBP-1
Applications Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin), Functional Assay
Classification Polyclonal
Concentration 1mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000, Flow Cytometry, Immunohistochemistry-Paraffin, Functional Assay
Formulation PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal
Gene Accession No. Q9BZM6
Gene Alias ALCAN-beta, N2DL1, N2DL-1, NKG2D ligand 1, NKG2DL1, RAET1Ialcan-beta, Retinoic acid early transcript 1I, UL16 binding protein 1, UL16-binding protein 1
Gene Symbols ULBP1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human ULBP1. The exact sequence is proprietary.
Molecular Weight of Antigen 28 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 80329
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.