Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ULBP-2/5/6 Antibody (165903) [DyLight 550], Novus Biologicals™
SDP

Catalog No. NB400443 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB400443 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB400443 Supplier Novus Biologicals Supplier No. FAB1298L
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ULBP-2/5/6 Monoclonal antibody specifically detects ULBP-2/5/6 in Human samples. It is validated for Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF

Specifications

Antigen ULBP-2/5/6
Applications Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF
Classification Monoclonal
Clone 165903
Conjugate DyLight 550
Dilution Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias ALCAN-alpha, N2DL2, NKG2DL2, RAET1H, RAET1L, UL16 binding protein 2, ULBP2
Host Species Mouse
Immunogen BaF3 mouse pro-B cell line transfected with human ULBP-2, Accession # Q9BZM5
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80328
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.