Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ULBP-2 Antibody (003), DyLight 350, Novus Biologicals™
SDP

Catalog No. NB069393 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB069393 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB069393 Supplier Novus Biologicals Supplier No. NBP290059UV
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

ULBP-2 Monoclonal specifically detects ULBP-2 in Human samples. It is validated for ELISA.

Specifications

Antigen ULBP-2
Applications ELISA
Classification Monoclonal
Clone 3
Conjugate DyLight 350
Dilution ELISA
Formulation 50mM Sodium Borate
Gene Alias ALCAN-alpha, N2DL2, N2DL-2, NKG2D ligand 2, RAET1HNKG2DL2, retinoic acid early transcript 1 H, Retinoic acid early transcript 1H, UL16 binding protein 2, UL16-binding protein 2
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human ULBP-2 (Q9BZM5; Met1-Ser217).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80328
Test Specificity No cross-reactivity in ELISA with: Human ULBP1
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.