Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ULBP-2 Antibody (003), Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. NB334760 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB334760 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB334760 Supplier Novus Biologicals Supplier No. NBP290059JF646
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

ULBP-2 Monoclonal antibody specifically detects ULBP-2 in Human samples. It is validated for ELISA

Specifications

Antigen ULBP-2
Applications ELISA
Classification Monoclonal
Clone 003
Conjugate Janelia Fluor 646
Dilution ELISA
Formulation 50mM Sodium Borate
Gene Alias ALCAN-alpha, N2DL2, N2DL-2, NKG2D ligand 2, RAET1HNKG2DL2, retinoic acid early transcript 1 H, Retinoic acid early transcript 1H, UL16 binding protein 2, UL16-binding protein 2
Host Species Rabbit
Immunogen Rabbit immunized with purified, recombinant Human ULBP-2 (Uniprot#: Q9BZM5, Met1-Ser217)
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 80328
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.