Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ULBP-2 Antibody (MM0593-7F33) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB018980 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB018980 0.025 mg
NBP211987 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB018980 Supplier Novus Biologicals Supplier No. NBP2119870.025MG

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse Monoclonal Antibody

ULBP-2 Monoclonal specifically detects ULBP-2 in Human samples. It is validated for Block/Neutralize.

Specifications

Antigen ULBP-2
Applications Blocking Assay
Classification Monoclonal
Clone MM0593-7F33
Concentration LYOPH
Conjugate Unconjugated
Dilution Blocking/Neutralizing 50-200 molar excess
Formulation A 0.2 um filtered solution in PBS with No Preservative
Gene Accession No. Q9BZM5
Gene Alias ALCAN-alpha, N2DL2, N2DL-2, NKG2D ligand 2, RAET1HNKG2DL2, retinoic acid early transcript 1 H, Retinoic acid early transcript 1H, UL16 binding protein 2, UL16-binding protein 2
Gene Symbols ULBP2
Host Species Mouse
Immunogen Human ULBP2
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80328
Reconstitution Reconstitute with 0.2 ml sterilized PBS to a final concentration of 0.5 mg/ml.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.