Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ULBP-4/RAET1E Antibody (RAET1E 50/3), FITC, Novus Biologicals™
SDP

Catalog No. NBP250267F Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP250267F 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP250267F Supplier Novus Biologicals Supplier No. NBP250267F
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ULBP-4/RAET1E Monoclonal specifically detects ULBP-4/RAET1E in Human samples. It is validated for Flow Cytometry, ELISA.

Specifications

Antigen ULBP-4/RAET1E
Applications Flow Cytometry, ELISA
Classification Monoclonal
Clone RAET1E 50/3
Conjugate FITC
Dilution Flow Cytometry, ELISA
Formulation PBS with 0.05% Sodium Azide
Gene Alias bA350J20.7, LETALRAE-1-like transcript 4, Lymphocyte effector toxicity activation ligand, MGC125309, N2DL4, N2DL-4, NKG2D ligand 4, NKG2DL4, RAET1E2, retinoic acid early transcript 1ERL-4, ULBP4MGC125308
Gene Symbols RAET1E
Host Species Mouse
Immunogen Recombinant His-tagged protein to human extra-cellular domain of ULBP-4/RAET1E, grown in E.coli
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 135250
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.