Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ulex Europaeus Lectin 1 Antibody, Novus Biologicals™
SDP

Catalog No. NB11013922 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB11013922 0.25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB11013922 Supplier Novus Biologicals Supplier No. NB11013922
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Ulex Europaeus Lectin 1 Polyclonal specifically detects Ulex Europaeus Lectin 1 in Plant samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen Ulex Europaeus Lectin 1
Applications Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:500, Immunohistochemistry-Paraffin 1:500, Immunohistochemistry-Frozen 1:500
Formulation Whole antisera, delipidized with 15mM Sodium Azide
Gene Alias Gorse or Furze Lectin, UEA-1; H(O) lectin 1; Anti H(O) lectin I
Host Species Rabbit
Immunogen Anti-Ulex Europaeus-I (UEA-I) Lectin is developed in rabbit using purified Ulex Europaeus-I lectin from gorse seeds as the immunogen.
Purification Method Unpurified
Quantity 0.25 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity The antibody may be used for studies of the vascular structures. It reacts specifically with UEA-I bound to human endothelial cells of normal and neoplastic blood and lymphatic vessels. Its uniform reaction with vessels enables differentation between vessels and artificial slits in the tissue.
Target Species Plant
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.