Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ULK1 Antibody, Novus Biologicals™
SDP

Catalog No. NB287498 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB287498 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB287498 Supplier Novus Biologicals Supplier No. NBP323466
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

ULK1 Monoclonal antibody specifically detects ULK1 in Human samples. It is validated for Western Blot

Specifications

Antigen ULK1
Applications Western Blot
Classification Monoclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000 - 1:2000
Formulation 50% Glycerol/PBS, 1% BSA
Gene Alias ATG1, ATG1 autophagy related 1 homolog, ATG1A, EC 2.7.11, EC 2.7.11.1, FLJ38455, FLJ46475, KIAA0722, serine/threonine-protein kinase ULK1, UNC51, unc-51 (C. elegans)-like kinase 1, Unc51.1, Unc-51-like kinase 1, unc-51-like kinase 1 (C. elegans)
Host Species Rabbit
Immunogen A phospho-peptide corresponding to human ULK1 [Ser757] (Uniprot#: O75385)
Purification Method Protein A purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Autophagy, mTOR Pathway, Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 8408
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.