Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ULK3 Antibody, mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB343650 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB343650 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB343650 Supplier Novus Biologicals Supplier No. NBP241243MFV500
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ULK3 Polyclonal antibody specifically detects ULK3 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen ULK3
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate mFluor Violet 500 SE
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias DKFZP434C131, EC 2.7.11.1, FLJ90566, serine/threonine-protein kinase ULK3, Unc-51-like kinase 3, unc-51-like kinase 3 (C. elegans)
Host Species Rabbit
Immunogen Antibody was raised against a 19 amino acid peptide near the center of human ULK3. The immunogen is located within amino acids 230 - 280 of ULK3. Amino Acid Squence: DLLQRLLERDPSRRISFQD
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 25989
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.