Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ULK3 Rabbit anti-Human, Mouse, Rat, Clone: 5Z7Z6, Novus Biologicals™
SDP

Catalog No. NB168335 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB168335 100 μg
NB168336 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB168335 Supplier Novus Biologicals Supplier No. NBP316622100UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

ULK3 Monoclonal antibody specifically detects ULK3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunofluorescence

Specifications

Antigen ULK3
Applications Western Blot, Immunofluorescence
Classification Monoclonal
Clone 5Z7Z6
Conjugate Unconjugated
Dilution Western Blot 1:500 - 1:2000, Immunocytochemistry/Immunofluorescence
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias DKFZP434C131, EC 2.7.11.1, FLJ90566, serine/threonine-protein kinase ULK3, Unc-51-like kinase 3, unc-51-like kinase 3 (C. elegans)
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 373-472 of human ULK3 (Q6PHR2).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 25989
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.