Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ULK4 Antibody, Novus Biologicals™
SDP

Catalog No. NBP120229 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mL
0.15 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP120229 0.15 mL
NB018404 0.05 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP120229 Supplier Novus Biologicals Supplier No. NBP120229
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 3 publications

ULK4 Polyclonal specifically detects ULK4 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen ULK4
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:200-1:2000, Immunohistochemistry 1:200-1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunocytochemistry
Formulation Lyophilized from whole antisera with No Preservative
Gene Accession No. Q96C45
Gene Alias DKFZp434E1822, EC 2.7.11.1, FAM7C1, FLJ20574, REC01035, serine/threonine-protein kinase ULK4, Unc-51-like kinase 4, unc-51-like kinase 4 (C. elegans)
Gene Symbols ULK4
Host Species Rabbit
Immunogen A synthetic peptide from human ULK4 conjugated to blue carrier protein has been used as the antigen. The peptide shares 95% identity with mouse sequence.
Purification Method Unpurified
Quantity 0.15 mL
Regulatory Status RUO
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 54986
Test Specificity ULK4.
Reconstitution Reconstitute in 150 ul of sterile water. Centrifuge to remove any insoluble material. Glycerol may be added (1:1) for additional stability. Please note the sample size is provided in reconstituted format.
Target Species Human, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.