Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ULK4 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 3 publications
$207.00 - $499.50
Specifications
Antigen | ULK4 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 1:200-1:2000, Immunohistochemistry 1:200-1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunocytochemistry |
Applications | Immunofluorescence, Immunohistochemistry, Immunocytochemistry, Western Blot |
Classification | Polyclonal |
Description
ULK4 Polyclonal specifically detects ULK4 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
ULK4 | |
Western Blot 1:200-1:2000, Immunohistochemistry 1:200-1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunocytochemistry | |
Polyclonal | |
Antisera | |
RUO | |
Mouse, Human | |
DKFZp434E1822, EC 2.7.11.1, FAM7C1, FLJ20574, REC01035, serine/threonine-protein kinase ULK4, Unc-51-like kinase 4, unc-51-like kinase 4 (C. elegans) | |
ULK4 | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
LYOPH | |
Immunofluorescence, Immunohistochemistry, Immunocytochemistry, Western Blot | |
Unconjugated | |
Rabbit | |
Protein Kinase | |
Q96C45 | |
54986 | |
IgG | |
Unpurified | |
ULK4. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title