Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Restek™ Ultra 5microm Bulk Packing Material

Click to view available options
Type:
Ultra Amino Bulk Packing
Ultra C1 Bulk Packing
Ultra C18 Bulk Packing
Ultra C4 Bulk Packing
Ultra C8 Bulk Packing
Ultra Cyano Bulk Packing
Description
- High quality materials
- Pore Size: 100 Angstrom
- Particle Size: 5μm
- Quantity: 10g/Bottle
Specifications
Specifications
Type | Ultra Amino Bulk Packing |
Packaging Quantity | 10 g/bottle |
Particle Size | 5 μm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction